Skip to Content
Merck
All Photos(1)

Key Documents

141720

Sigma-Aldrich

Cycloheptene

97%

Synonym(s):

(1Z)-Cycloheptene, cis-Cycloheptene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12
CAS Number:
Molecular Weight:
96.17
Beilstein:
1900884
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.458 (lit.)

bp

112-114.7 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

SMILES string

C1CCC=CCC1

InChI

1S/C7H12/c1-2-4-6-7-5-3-1/h1-2H,3-7H2

InChI key

ZXIJMRYMVAMXQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of gas-phase reactions of cycloheptene with NO3 radicals and O3 has been investigated.

Application

Cycloheptene was used in one-pot synthesis of 1,2/3-triols from the allylichydroperoxides catalyzed by zeolite-confined osmium(0) nanoclusters.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-pot synthesis of 1, 2/3-triols from the allylic hydroperoxides catalyzed by zeolite-confined osmium (0) nanoclusters.
Goksu H, et al.
J. Mol. Catal. A: Chem., 378, 142-147 (2013)
Alla Zelenyuk et al.
Faraday discussions, 200, 143-164 (2017-06-06)
When secondary organic aerosol (SOA) particles are formed by ozonolysis in the presence of gas-phase polycyclic aromatic hydrocarbons (PAHs), their formation and properties are significantly different from SOA particles formed without PAHs. For all SOA precursors and all PAHs, discussed
Sherri A Mason et al.
The journal of physical chemistry. A, 113(19), 5649-5656 (2009-04-24)
Rate constants for the gas-phase reactions of NO(3) radicals and O(3) with a series of C(6)-C(14) 1-alkenes and 2-methyl-1-alkenes have been measured at 296 +/- 2 K and atmospheric pressure of air using relative rate methods. For the NO(3) radical
Jong-Ryul Park et al.
Biointerphases, 16(1), 011001-011001 (2021-01-07)
Poly(2-alkyl-2-oxazoline) (PAOx) hydrogels are tailorable synthetic materials with demonstrated biomedical applications, thanks to their excellent biocompatibility and tunable properties. However, their use as injectable hydrogels is challenging as it requires invasive surgical procedures to insert the formed hydrogel into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service