Skip to Content
Merck
All Photos(1)

Documents

133108

Sigma-Aldrich

Propyl acetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2CH3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740764
EC Number:
MDL number:
UNSPSC Code:
12352108
eCl@ss:
39022103
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.5 (vs air)

Quality Level

vapor pressure

25 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

842 °F

expl. lim.

1.7 %, 37 °F
8 %

refractive index

n20/D 1.384 (lit.)

bp

102 °C (lit.)

mp

−95 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

CCCOC(C)=O

InChI

1S/C5H10O2/c1-3-4-7-5(2)6/h3-4H2,1-2H3

InChI key

YKYONYBAUNKHLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Identification and biotransformation analysis of volatile markers during the early stage of Salmonella contamination in chicken.: This study highlights the role of propyl acetate in identifying volatile organic compounds as markers for early Salmonella contamination in poultry products, using sophisticated analytical techniques to ensure food safety (Wang et al., 2024).


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.2 °F - closed cup

Flash Point(C)

11.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Greening the esterification between isosorbide and acetic acid
Inayat A, et al.
Sustainable Chemistry and Pharmacy, 7, 41-49 (2018)
Extraction equilibria of 2, 4, 6-Trinitrophenol by (amberlite LA2+ ester) solvents
Uslu H
Fluid Phase Equilibria, 427, 175-179 (2016)
Feasibility of reactive chromatography for the synthesis of n-propyl acetate
Reddy B and Mahajani S
Industrial & Engineering Chemistry Research, 53(4), 1395-1403 (2014)
Liquid-liquid equilibria for the ternary system water+ acetic acid+ propyl acetate.
Xiao X, et al.
Journal of Chemical and Engineering Data, 51(2), 582-583 (2006)
High polymer solutions. Part 7.-Vapour pressure of polystyrene solutions in acetone, chloroform and propyl acetate.
Bawn CEH and Wajid MA.
Transactions of the Faraday Society, 52, 1658-1664 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service