Skip to Content
Merck
All Photos(1)

Key Documents

128422

Sigma-Aldrich

mono-Ethyl fumarate

95%

Synonym(s):

Fumaric acid monoethyl ester, Monoethyl fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH=CHCOOH
CAS Number:
Molecular Weight:
144.13
Beilstein:
1723588
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

147 °C/16 mmHg (lit.)

mp

66-68 °C (lit.)

SMILES string

CCOC(=O)\C=C\C(O)=O

InChI

1S/C6H8O4/c1-2-10-6(9)4-3-5(7)8/h3-4H,2H2,1H3,(H,7,8)/b4-3+

InChI key

XLYMOEINVGRTEX-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

mono-Ethyl fumarate (fumaric acid monoethyl ester, monoethyl fumarate) was used in the preparation of photo-crosslinkable macromers. It was also used to synthesize Ugi/intramolecular Diels-Alder (IMDA) cycloaddition products.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Paulvannan
The Journal of organic chemistry, 69(4), 1207-1214 (2004-02-14)
An efficient approach to rigid tricyclic nitrogen heterocycles via sequential and tandem Ugi/intramolecular Diels-Alder (IMDA) cycloaddition of pyrrole is described. The one-pot Ugi four-component condensation (4CC) reaction was used as the key transformation to prepare trienes with a carboxamide substituent
Janine Jansen et al.
Acta biomaterialia, 8(10), 3652-3659 (2012-06-19)
Photo-crosslinked networks were prepared from fumaric acid monoethyl ester-functionalized poly(D,L-lactic acid) oligomers and N-vinyl-2-pyrrolidone. Two model proteins, lysozyme and albumin, were incorporated into the network films as solid particles and their release behavior was studied. By varying the NVP content
J Z Hu et al.
Solid state nuclear magnetic resonance, 6(1), 85-94 (1996-02-01)
The 1H magic-angle spinning (MAS) spectrum for a typical powdered solid is composed of a high resolution component and a broadline component. The high resolution component can be well isolated from the broadline component by the Hahn echo sequence with
B Sebök et al.
European journal of pharmacology, 270(1), 79-87 (1994-01-03)
Oral administration with complex mixtures of fumaric acid derivatives is known to have antipsoriatic efficacy. The present studies aimed to clarify the mode of action and toxicity of the individual compounds. Hyperproliferative HaCaT keratinocytes in monolayer cultures were exposed to
Treatment of recurrent aphthous stomatitis with fumaric acid esters.
Emmanuella Guenova et al.
Archives of dermatology, 147(3), 282-284 (2011-03-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service