Skip to Content
Merck
All Photos(3)

Key Documents

127744

Sigma-Aldrich

Diethylamine hydrochloride

ReagentPlus®, 99%

Synonym(s):

Diethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH · HCl
CAS Number:
Molecular Weight:
109.60
Beilstein:
3590084
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

form

solid

mp

227-230 °C (lit.)

solubility

H2O: soluble 50 mg/mL, clear, colorless
H2O: soluble, clear, colorless (50 mg/mL)

SMILES string

Cl.CCNCC

InChI

1S/C4H11N.ClH/c1-3-5-4-2;/h5H,3-4H2,1-2H3;1H

InChI key

HDITUCONWLWUJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylamine hydrochloride has been used in the synthesis of diethylaminoethyl (DEAE) cottons by reacting it with cotton cellulose in the presence of sodium hydroxide.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hebeish A and El-Hilw ZH.
Journal of Applied Polymer Science, 67(4) (1998)
Min-Jon Lin et al.
Biochemical and biophysical research communications, 365(4), 724-728 (2007-11-24)
In this study, we investigated the effect of NO donor, diethylamine/nitric oxide (DEA/NO), on the electrophysiological behavior of human skeletal muscle chloride channel (CLCN1). The wild-type and variants of CLCN1, including one polymorphism (P727L) and four mutants (T631I, D644G, G482R
Jingfeng Chen et al.
Biochimica et biophysica acta, 1807(5), 491-502 (2011-03-17)
Mitochondria-derived oxygen-free radical(s) are important mediators of oxidative cellular injury. It is widely hypothesized that excess NO enhances O(2)(•-) generated by mitochondria under certain pathological conditions. In the mitochondrial electron transport chain, succinate-cytochrome c reductase (SCR) catalyzes the electron transfer
Helmut Görner
The journal of physical chemistry. A, 115(29), 8208-8215 (2011-06-22)
The photoreduction of 6-nitrospiro[2H-1-benzopyran-2,2'-indoline] (N1) and two derivatives (N2 and N3) by diethylamine or triethylamine (TEA) in solution was studied by pulsed and steady-state photolysis. The quantum yield of coloration of the ring-closed Sp form, due to photoinduced ring opening
Nathalie Hasler-Nguyen et al.
BMC research notes, 5, 321-321 (2012-06-23)
Rubbing a topical NSAID (non steroidal anti-inflammatory drug) on the skin may increase local drug permeation, affecting its distribution to the site of pain and inflammation. The present study evaluates this hypothesis, by assessing in vitro the effect on skin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service