Skip to Content
Merck
All Photos(1)

Key Documents

127698

Sigma-Aldrich

2-Nitrobenzoic acid

95%, Contains 3- and 4-isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CO2H
CAS Number:
Molecular Weight:
167.12
Beilstein:
1910632
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

powder

mp

146-148 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1ccccc1[N+]([O-])=O

InChI

1S/C7H5NO4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)

InChI key

SLAMLWHELXOEJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Nitrobenzoic acid can function as a growth supplement for Pseudomonas fluorescens strain KU-7[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Hasegawa et al.
FEMS microbiology letters, 190(2), 185-190 (2000-10-18)
A bacterial strain KU-7, identified as a Pseudomonas fluorescens by 16S rDNA sequencing, was one of the 12 new isolates that are able to grow on 2-nitrobenzoate as a sole source of carbon, nitrogen, and energy. Resting cells of KU-7
Lei Wang et al.
Bioconjugate chemistry, 25(7), 1252-1260 (2014-06-28)
A new strategy for accurate and reversible modulation of protein activity via simple conjugation of the sulfhydryl modifier and polymer with the introduced Cys residue in protein was developed in this study. With Escherichia coli inorganic pyrophosphatase (PPase) as a
Jing Yang et al.
Journal of biophotonics, 8(8), 659-667 (2014-09-27)
We experimentally demonstrate an ultra-sensitive immunoassay biosensor using diatom biosilica with self-assembled plasmonic nanoparticles. As the nature-created photonic crystal structures, diatoms have been adopted to enhance surface plasmon resonances of metal nanoparticles on the surfaces of diatom frustules and to
P A Ferchmin et al.
Neurotoxicology, 44, 80-90 (2014-06-15)
Many organophosphorous esters synthesized for applications in industry, agriculture, or warfare irreversibly inhibit acetylcholinesterase, and acute poisoning with these compounds causes life-threatening cholinergic overstimulation. Following classical emergency treatment with atropine, an oxime, and a benzodiazepine, surviving victims often suffer brain
Rashmi Tupe et al.
International journal of biological macromolecules, 79, 601-610 (2015-06-02)
The present work aims to investigate the concentration and time dependant effect of zinc on the in vitro non enzymatic modifications of albumin by diabetic levels of glucose. Further, preventive and curative effect of zinc was studied by adding zinc

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service