Skip to Content
Merck
All Photos(2)

Documents

125199

Sigma-Aldrich

3-Chloropropanesulfonyl chloride

98%

Synonym(s):

γ-Chloropropanesulfonyl chloride, 1-Chloro-3-propanesulfonyl chloride, 3-Chloro-1-propanesulfonyl chloride, 3-Chloro-n-propanesulfonyl chloride, 3-Chloropropylsulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3SO2Cl
CAS Number:
Molecular Weight:
177.05
Beilstein:
1754119
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.489 (lit.)

bp

70 °C/0.5 mmHg (lit.)

density

1.456 g/mL at 25 °C (lit.)

SMILES string

ClCCCS(Cl)(=O)=O

InChI

1S/C3H6Cl2O2S/c4-2-1-3-8(5,6)7/h1-3H2

InChI key

GPKDGVXBXQTHRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chloropropanesulfonyl chloride was used as starting reagent in the synthesis of the pyrrolidine-based chiral imidazolium ionic liquid. It was also used in sulfonation of cross-linked polyethylenimine for selective removal of Hg. It was used in generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids to yield chiral sultams and sultones.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic enantio- and diastereoselective formation of beta-sultones: ring-strained precursors for enantioenriched beta-hydroxysulfonyl derivatives.
Florian M Koch et al.
Angewandte Chemie (International ed. in English), 46(15), 2685-2689 (2007-03-03)
Functionalized chiral ionic liquid as recyclable organocatalyst for asymmetric Michael addition to nitrostyrenes.
Ni B, et al.
Green Chemistry, 9(7), 737-739 (2007)
Sulfonated cross-linked polyethylenimine for selective removal of mercury from aqueous solutions.
Saad DMG, et al.
Toxicological & Environmental Chemistry, 94(10), 1916-1929 (2012)
Xolisiwe Sebutsoe et al.
Chemosphere, 266, 128975-128975 (2020-11-25)
Arsenic and selenium are both carcinogenic and their presence in fresh water has attracted the development of robust and accurate monitoring techniques. A new diffusive gradients in thin-films (DGT) sampler was developed and evaluated for the in situ measurements of
Marian Zajac et al.
Organic letters, 9(10), 2007-2010 (2007-04-24)
beta-Sultams, highly strained sulfonyl analogues of beta-lactams, were prepared enantio- and diastereoselectively by tertiary amine catalyzed [2 + 2] cycloaddition reactions. The title compounds are practical precursors of highly enantioenriched biologically interesting beta-aminosulfonyl derivatives.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service