Skip to Content
Merck
All Photos(1)

Key Documents

11373

Sigma-Aldrich

HATU

≥98.0% (CHN), for peptide synthesis

Synonym(s):

1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate, N-[(Dimethylamino)-1H-1,2,3-triazolo-[4,5-b]pyridin-1-ylmethylene]-N-methylmethanaminium hexafluorophosphate N-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15F6N6OP
CAS Number:
Molecular Weight:
380.23
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Product Name

HATU, ≥98.0% (CHN)

Assay

≥98.0% (CHN)

form

solid

reaction suitability

reaction type: Coupling Reactions

mp

183-185 °C (lit.)
183-188 °C (dec.)

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)[C+](N(C)C)n1n[n+]([O-])c2ncccc12

InChI

1S/C10H15N6O.F6P/c1-13(2)10(14(3)4)15-8-6-5-7-11-9(8)16(17)12-15;1-7(2,3,4,5)6/h5-7H,1-4H3;/q+1;-1

InChI key

FKBFHOSFPRWJNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for:
Synthesis of Aurora A kinase inhibitors
HPLC assay to determine D- and L- acid enantiomers in human plasma
Amide bond formation reactions

Catalyst for:
Selective acylation
Selecocyclization-oxidation deselenation sequence

Other Notes

Recent studies show that crystal as well as solution structure of this coupling agent is a guanidinium N-oxide and not an uronium compound; Advantageous coupling reagent for solid-phase peptide synthesis.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1A

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The uronium/guanidinium Peptide coupling reagents: finally the true uronium salts.
Louis A Carpino et al.
Angewandte Chemie (International ed. in English), 41(3), 441-445 (2002-12-20)
Qingxin Chen et al.
Journal of virology, 94(19) (2020-07-17)
Adeno-associated virus (AAV) has proven to be a promising candidate for gene therapy due to its nonpathogenic nature, ease of production, and broad tissue tropism. However, its transduction capabilities are not optimal due to the interaction with various host factors
L. Carpino et al.
Journal of the Chemical Society. Chemical Communications, 201-201 (1994)
Mazen Jamous et al.
Nuclear medicine and biology, 41(6), 464-470 (2014-05-02)
The gastrin-releasing peptide receptor (GRPR) was shown to be expressed with high density on several types of cancers. Radiolabeled peptides for imaging and targeted radionuclide therapy have been developed. In this study, we evaluated the potential of statine-based bombesin antagonists
Anna K Lee et al.
Molecular cell, 79(4), 645-659 (2020-07-22)
Stress granules (SGs) are membrane-less ribonucleoprotein condensates that form in response to various stress stimuli via phase separation. SGs act as a protective mechanism to cope with acute stress, but persistent SGs have cytotoxic effects that are associated with several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service