Skip to Content
Merck
All Photos(2)

Key Documents

112577

Sigma-Aldrich

L-(−)-Malic acid

97%, optical purity ee: 99% (GLC)

Synonym(s):

(S)-(−)-2-Hydroxysuccinic acid, L-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723541
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

powder

optical activity

[α]20/D −27°, c = 5.5 in pyridine

optical purity

ee: 99% (GLC)

mp

101-103 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

O[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m0/s1

InChI key

BJEPYKJPYRNKOW-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-(-)-Malic acid is an organic acid that is commonly found in wine. It plays an important role in wine microbiological stability.[1]

Application

L-(-)-Malic acid is a selective α-amino protecting reagent for amino acid derivatives.[2] It is also a versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists,[3] 1α,25-dihydroxyvitamin D3 analogue,[4] and phoslactomycin B.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic determination of L (-) malic acid in wines using sequential injection analysis.
Segundo MA and Rangel AOSS.
Analytica Chimica Acta, 499(1), 99-106 (2003)
Bioorganic & Medicinal Chemistry Letters, 2, 1713-1713 (1992)
Zhigen Li et al.
Journal of synchrotron radiation, 27(Pt 1), 100-109 (2019-12-24)
Aluminium (Al) K- and L-edge X-ray absorption near-edge structure (XANES) has been used to examine Al speciation in minerals but it remains unclear whether it is suitable for in situ analyses of Al speciation within plants. The XANES analyses for
I Yoshpe-Besançon et al.
Biotechnology and applied biochemistry, 18 ( Pt 1), 93-102 (1993-08-01)
In previous papers we have reported that an aminopeptidase A (EC 3.4.11.7) purified from Staphylococcus chromogenes was able to catalyse the introduction of L-malic acid at the N-terminus of Tyr and Phe derivatives. We now show that this enzyme can
Tetrahedron Letters, 48, 5601-5601 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service