Skip to Content
Merck
All Photos(2)

Key Documents

109975

Sigma-Aldrich

1,2-Epoxybutane

99%

Synonym(s):

α-Butylene oxide, 1,2-Butylene oxide, 1-Butene oxide, Ethyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102411
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

2.2 (vs air)

vapor pressure

140 mmHg ( 20 °C)

Assay

99%

autoignition temp.

698 °F

expl. lim.

19 %

refractive index

n20/D 1.384 (lit.)

bp

63 °C (lit.)

density

0.829 g/mL at 25 °C (lit.)

SMILES string

CCC1CO1

InChI

1S/C4H8O/c1-2-4-3-5-4/h4H,2-3H2,1H3

InChI key

RBACIKXCRWGCBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Epoxybutane can be used:
  • As a monomer to synthesize novel initiators via ring-opening polymerization. These initiators can be used to prepare complex macromolecules such as grafted polyamides.
  • To functionalize polyethyleneimine which is used in the synthesis of oxidation-stable adsorbents for CO2 capture.

Features and Benefits

  • High polymerizability
  • Low susceptibility to transfer reaction
  • Ease of handling

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengyuan Zhou et al.
International journal of pharmaceutics, 354(1-2), 82-87 (2007-12-08)
Ethylene oxide and 1,2-butylene oxide were sequentially polymerised to form the diblock copolymer E13B10 (E=oxyethylene, B=oxybutylene, subscripts denote number-average block lengths in repeat units). Dynamic and static light scattering over the temperature range 10-30 degrees C demonstrated a transition from
D Weinstein et al.
Environmental mutagenesis, 3(1), 1-9 (1981-01-01)
Based on the findings of Nagao et al [1978] that phenacetin is negative in the standard Ames test with Aroclor induced rat S-9 and positive with hamster S-9, the test was performed with a mixture of rat/hamster S-9. Phenacetin was
D Henschler et al.
Journal of cancer research and clinical oncology, 107(3), 149-156 (1984-01-01)
Previous analytical studies of industrial samples of trichloroethylene (TRI) have revealed the presence of mutagenic and carcinogenic epoxides which, it was proposed, might be responsible for the carcinogenicity of such samples, as demonstrated with mice in other laboratories. To test
A Ohnishi et al.
Environmental research, 60(2), 242-247 (1993-02-01)
Axonal neuropathy occurs due to occupational ethylene oxide (EtO) exposure. The experimental model of human EtO neuropathy was established. In addition, the neurotoxic effects of propylene oxide (PpO) and butylene oxide (BtO) were demonstrated in rats. Although no human neuropathy
Quantitative determination of 1,2-butylene oxide in whole rat blood by gas chromatography-mass spectrometry.
P E Kastl et al.
Journal of chromatography, 398, 347-350 (1987-07-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service