Skip to Content
Merck
All Photos(2)

Key Documents

107646

Sigma-Aldrich

Tripentaerythritol

technical grade

Synonym(s):

2,2-Bis[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-1,3-propanediol, 2-[[3-Hydroxy-2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)3CCH2OCH2C(CH2OH)2CH2OCH2C(CH2OH)3
CAS Number:
Molecular Weight:
372.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

solid

mp

225 °C (dec.) (lit.)

functional group

ether
hydroxyl

SMILES string

OCC(CO)(CO)COCC(CO)(CO)COCC(CO)(CO)CO

InChI

1S/C15H32O10/c16-1-13(2-17,3-18)9-24-11-15(7-22,8-23)12-25-10-14(4-19,5-20)6-21/h16-23H,1-12H2

InChI key

PTJWCLYPVFJWMP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study on the Preparation of Tripentaerythritol Phosphite Intumescent Flame Retardant and Its Application in Epoxy Resins.
Chun-mei S, et al.
Journal of Functional Polymers / Gong Neng Gao Fen Zi Xue Bao, 18(2), 260-260 (2005)
Jenna Usprech et al.
Acta biomaterialia, 58, 34-43 (2017-05-24)
The physical and chemical properties of a biomaterial integrate with soluble cues in the cell microenvironment to direct cell fate and function. Predictable biomaterial-based control of integrated cell responses has been investigated with two-dimensional (2D) screening platforms, but integrated responses
Study on intumescent flame retarded polystyrene composites with improved flame retardancy.
Lu H and Wilkie CA.
Polymer Degradation and Stability, 95(12), 2388-2395 (2010)
Blair J Sampson et al.
Journal of economic entomology, 112(1), 173-180 (2018-10-03)
Spotted-wing Drosophila, Drosophila suzukii (Matsumura) (Diptera: Drosophilidae) is a vinegar fly introduced unintentionally into the United States. Since 2008, D. suzukii has reduced annual berry yields from 6 to 100%. Effective control of D. suzukii during harvest requires weekly applications
Zhuojun Huang et al.
Biomaterials science, 6(9), 2487-2495 (2018-08-03)
We report here the development of hydrogels formed at physiological conditions using PEG (polyethylene glycol) based polymers modified with boronic acids (BAs) as backbones and the plant derived polyphenols ellagic acid (EA), epigallocatechin gallate (EGCG), tannic acid (TA), nordihydroguaiaretic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service