Skip to Content
Merck
All Photos(3)

Key Documents

104051

Sigma-Aldrich

Picolinamide

98%

Synonym(s):

2-Pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

110 °C (dec.) (lit.)

functional group

amide

SMILES string

NC(=O)c1ccccn1

InChI

1S/C6H6N2O/c7-6(9)5-3-1-2-4-8-5/h1-4H,(H2,7,9)

InChI key

IBBMAWULFFBRKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Picolinamide was used as template in preparation of molecular imprinting polymer. Picolinamide was used in a study to evaluate kinetics and mechanism of liberation of picolinamide from chromium(III)-picolinamide complexes in HClO4.

Biochem/physiol Actions

Picolinamide is potential inhibitor of poly (ADP-ribose) synthetase of nuclei from rat pancreatic islet cells. Picolinamide acts as bidentate ligand and forms complexes with lanthanide nitrates, thiocyanates and perchlorates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuela Jörg et al.
ChemMedChem, 16(1), 216-233 (2020-08-28)
This study investigated the structure-activity relationships of 4-phenylpyridin-2-one and 6-phenylpyrimidin-4-one M1 muscarinic acetylcholine receptor (M1 mAChRs) positive allosteric modulators (PAMs). The presented series focuses on modifications to the core and top motif of the reported leads, MIPS1650 (1) and MIPS1780
Mark Turlington et al.
Journal of medicinal chemistry, 56(20), 7976-7996 (2013-09-21)
Positive allosteric modulators (PAMs) of metabotropic glutamate receptor 5 (mGlu5) represent a promising therapeutic strategy for the treatment of schizophrenia. Both allosteric agonism and high glutamate fold-shift have been implicated in the neurotoxic profile of some mGlu5 PAMs; however, these
Hongguang Liu et al.
Journal of medicinal chemistry, 56(3), 895-901 (2013-01-11)
Melanoma is an aggressive skin cancer with worldwide increasing incidence. Development of positron emission tomography (PET) probes for early detection of melanoma is critical for improving the survival rate of melanoma patients. In this research, (18)F-picolinamide-based PET probes were prepared
T Kawabata et al.
Acta pathologica japonica, 42(7), 469-475 (1992-07-01)
The effects of three isomers of pyridinecarboxamide (picolinamide (2-pyridinecarboxamide), nicotinamide (3-pyridinecarboxamide) and isonicotinamide (4-pyridinecarboxamide)) on iron-induced renal damage were studied. Pyridinecarboxamide (250 mg/kg body weight, ip) was administered 10 min before injection of ferric nitrilotriacetate (Fe(III)-NTA) (7.5 mgFe/kg body weight
Lanthanide complexes with picolinamide.
Condorelli G, et al.
J. Inorg. Nucl. Chem., 36(12), 3763-3766 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service