Skip to Content
Merck
All Photos(1)

Key Documents

102741

Sigma-Aldrich

2-Fluorobiphenyl

96%

Synonym(s):

1-Fluoro-2-phenylbenzene, o-Fluorodiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4F
CAS Number:
Molecular Weight:
172.20
Beilstein:
2043175
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

solid

bp

248 °C (lit.)

mp

71-74 °C (lit.)

functional group

fluoro
phenyl

SMILES string

Fc1ccccc1-c2ccccc2

InChI

1S/C12H9F/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H

InChI key

KLECYOQFQXJYBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Fluorobiphenyl is a white to off white powder. It is catabolized by C. elegans, yielding hydroxylated products[1]. The metabolism of 2-fluorobiphenyl by hepatic microsomes isolated from control and aroclor 1254-treated rats and pigeons was studied[2].

Application

2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry[3]. It was used to study the solvent dynamics of 1,2,4,5-tetracyanobenzene/biphenyl derivative radical ion pairs by picosecond absorption spectroscopy[4]. It was also used as a sole carbon and energy source for the biphenyl-degrading bacterium Pseudomonas pseudoalcaligenes KF707[5].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cormac D Murphy et al.
FEMS microbiology letters, 286(1), 45-49 (2008-07-12)
The biphenyl-degrading bacterium Pseudomonas pseudoalcaligenes KF707 can use 2- and 4-fluorobiphenyl as sole carbon and energy sources. Accumulation of fluorinated catabolites was determined by fluorine-19 nuclear magnetic spectroscopy (19F NMR) and revealed that growth on 4-fluorobiphenyl yielded 4-fluorobenzoate and 4-fluoro-1,2-dihydro-1,2-dihydroxybenzoate
Jessica Amadio et al.
Applied microbiology and biotechnology, 86(1), 345-351 (2009-12-04)
The fungus Cunninghamella elegans is a useful model of human catabolism of xenobiotics. In this paper, the biotransformation of fluorinated biphenyls by C. elegans was investigated by analysis of the culture supernatants with a variety of analytical techniques. 4-Fluorobiphenyl was
Zheng-Xia Chen et al.
Chemosphere, 131, 192-200 (2015-04-12)
Fine roots of woody plants are a heterogeneous system differing markedly in structure and function. Nevertheless, knowledge about the plant uptake of organic pollutants via fine roots is scarce to date. In the present study, plant uptake, translocation, and return
Solvent Effect on the Dynamics of Radical Ion Pair Separation.
Han CH.
Journal of Photoscience, 8(1), 33-37 (2001)
Coriness Piñeyro-Ruiz et al.
Frontiers in pediatrics, 8, 202-202 (2020-05-12)
Background & Objective: Hypospadias, characterized by the displacement of the opening of the urethra at any point in the medial-ventral side of the penis, is classified upon severity as mild (Type I) and severe (Type II and Type III) hypospadias.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service