Skip to Content
Merck
All Photos(1)

Key Documents

101370

Sigma-Aldrich

2-Methyl-1-undecene

97%

Synonym(s):

1-Methyl-1-nonylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8C(CH3)=CH2
CAS Number:
Molecular Weight:
168.32
Beilstein:
1739514
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.432 (lit.)

density

0.757 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC(C)=C

InChI

1S/C12H24/c1-4-5-6-7-8-9-10-11-12(2)3/h2,4-11H2,1,3H3

InChI key

SJVKHZYVCVKEGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Lommatzsch et al.
Journal of chromatography. A, 1402, 94-101 (2015-06-04)
Multidimensional chromatography based on two-dimensional high performance liquid chromatography on-line coupled to gas chromatography (on-line HPLC-HPLC-GC) enables the separate analysis of saturated, monounsaturated and aromatic hydrocarbons in packaging materials like polyolefins or paperboard and their migrates into foods. Since normal-phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service