Merck
  • The actin binding cytoskeletal protein Moesin is involved in nuclear mRNA export.
The actin binding cytoskeletal protein Moesin is involved in nuclear mRNA export.
Biochimica et biophysica acta. Molecular cell research (2017-05-31)
Ildikó Kristó, Csaba Bajusz, Barbara N Borsos, Tibor Pankotai, Joseph Dopie, Ferenc Jankovics, Maria K Vartiainen, Miklós Erdélyi, Péter Vilmos
ABSTRACT
Current models imply that the evolutionarily conserved, actin-binding Ezrin-Radixin-Moesin (ERM) proteins perform their activities at the plasma membrane by anchoring membrane proteins to the cortical actin network. Here we show that beside its cytoplasmic functions, the single ERM protein of Drosophila, Moesin, has a novel role in the nucleus. The activation of transcription by heat shock or hormonal treatment increases the amount of nuclear Moesin, indicating biological function for the protein in the nucleus. The distribution of Moesin in the nucleus suggests a function in transcription and the depletion of mRNA export factors Nup98 or its interacting partner, Rae1, leads to the nuclear accumulation of Moesin, suggesting that the nuclear function of the protein is linked to mRNA export. Moesin localizes to mRNP particles through the interaction with the mRNA export factor PCID2 and knock down of Moesin leads to the accumulation of mRNA in the nucleus. Based on our results we propose that, beyond its well-known, manifold functions in the cytoplasm, the ERM protein of Drosophila is a new, functional component of the nucleus where it participates in mRNA export.
Sigma-Aldrich
Triptolide, from Tripterygium wilfordii, ≥98% (HPLC), solid
Sigma-Aldrich
5-Fluorouridine, proapoptotic anitproliferative plant growth regulator
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Triptolide, Tripterygium wilfordii, A novel diterpene triepoxide isolated from the Chinese herb Tripterygium wilfordii that acts as a potent immunosuppressant and anti-inflammatory agent.
Sigma-Aldrich
Anti-HA tag antibody produced in chicken, affinity isolated antibody
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid