Merck
  • Evidence against a role for β-arrestin1 in STAT1 dephosphorylation and the inhibition of interferon-γ signaling.
Evidence against a role for β-arrestin1 in STAT1 dephosphorylation and the inhibition of interferon-γ signaling.
Molecular cell (2013-04-16)
Christin Pelzel, Andreas Begitt, Nikola Wenta, Uwe Vinkemeier
ABSTRACT
Signal transducer and activator of transcription 1 (STAT1) is activated by tyrosine phosphorylation upon interferon-γ (IFNγ) stimulation, which results in the expression of genes with antiproliferative and immunomodulatory functions. The inactivation of STAT1 occurs through tyrosine dephosphorylation by the tyrosine phosphatase TC45. It was proposed that recruitment of TC45 required the direct interaction of STAT1 with the scaffold protein β-arrestin1, making β-arrestin1 an essential negative regulator of STAT1 and IFNγ signaling (Mo et al., 2008). We tested the relevance of β-arrestin1 for STAT1 activity. Our results do not confirm β-arrestin1 as a STAT1-interacting protein. The STAT1 phosphorylation/dephosphorylation cycle was found to be unaffected by both the overexpression and the genetic deletion of β-arrestin1. Accordingly, β-arrestin1 did not inhibit STAT1 transcriptional activity or the induction of IFNγ target genes in response to IFNγ. Our data indicate that β-arrestin1 is dispensable for STAT1 dephosphorylation and the termination of IFNγ signaling.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
γ-Interferon, Human, Recombinant, E. coli
Sigma-Aldrich
γ-Interferon, Mouse, Recombinant, E. coli
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Staurosporine, Streptomyces sp., Staurosporine, CAS 62996-74-, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinases (IC₅₀ = 7, 20, 1.3, 0.7, & 8.5 nM for PKA, CAMK, MLCK, PKC, & PKG, respectively).