Merck
  • Derivatization of bisphenol A and its analogues with pyridine-3-sulfonyl chloride: multivariate optimization and fragmentation patterns by liquid chromatography/Orbitrap mass spectrometry.
Derivatization of bisphenol A and its analogues with pyridine-3-sulfonyl chloride: multivariate optimization and fragmentation patterns by liquid chromatography/Orbitrap mass spectrometry.
Rapid communications in mass spectrometry : RCM (2015-07-28)
Jorge Regueiro, Andreas Breidbach, Thomas Wenzl
ABSTRACT
Due to the growing restrictions on the use of bisphenol A (BPA), several other bisphenols are gaining importance as substitutes for BPA in a variety of applications. There is, therefore, a real need for selective and sensitive methods based on mass spectrometry which will allow the human exposure to these new bisphenols to be assessed. Derivatization of BPA and its substitutes with pyridine-3-sulfonyl chloride is used to enhance the detection capability of bisphenols by electrospray ionization mass spectrometry. A multivariate experimental design, Box-Behnken response surface, was used to evaluate the influence of the main variables potentially affecting the derivatization yield. Fragmentation patterns for all the derivatized bisphenols were acquired by high-resolution/accurate-mass Orbitrap mass spectrometry. Temperature and pH were identified as the most important factors affecting the derivatization yield of bisphenols. Fragmentation of the protonated molecules produced abundant analyte-specific product ions. Most of the derivatized bisphenols showed significant improvements in their signal-to-noise ratios compared with the underivatized forms. The stability of these derivatives was demonstrated through several freeze/thaw cycles, short-term room temperature and long-term cold storage. Derivatization of BPA and its structural analogues with pyridine-3-sulfonyl chloride is proposed as a specific, sensitive, high-throughput approach to their analysis by liquid chromatography coupled to electrospray ionization mass spectrometry.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
3-pyridinesulfonyl chloride hydrochloride, AldrichCPR
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
4,4′-(1-Phenylethylidene)bisphenol, 99%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
4,4′-(Hexafluoroisopropylidene)diphenol, 97%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
4,4′-Cyclohexylidenebisphenol, 98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG