Skip to Content
Merck
All Photos(1)

Key Documents

H9892

Sigma-Aldrich

Hydrochloric acid

1.0 N, BioReagent, suitable for cell culture

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.71

Product Name

Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture

grade

reagent

sterility

sterile-filtered

product line

BioReagent

form

solution

concentration

1.0 N

technique(s)

cell culture | mammalian: suitable

anion traces

chloride (Cl-): passes test

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrochloric acid solution, a clear colorless liquid, is a solution of hydrogen chloride gas dissolved in water. It is a strong inorganic acid.

Application

Hydrochloric acid solution (HCl solution) is mainly used to adjust the pH in cell culture medium.
It can also be used:
  • In the extraction of elderberry anthocyanins by endothelial cells.
  • To convert iron(III) perchlorate to Fe(III) ions which can then form a complex with acetohydroxamic acid (FeAHA).

The hydrochloric acid solution has been used:
  • to acidify the surface water sample and to prevent interference from carbonate concentrations up to 10 mg L-1 with organic carbon detector (OCD) detection
  • to adjust the pH of complete transfection media for transfection with chitosan/pDNA nanoparticle complexes
  • to prepare artificial tear solution (ATS) to incorporate the fluorescently tagged cholesteryl ester

Biochem/physiol Actions

Hydrochloric acid (HCl) is commonly used in research and large-scale applications. Its large-scale applications include the refinement of ore to produce tin and tantalum, the cleaning of metal products, and the hydrolysis of starch and proteins in the production of food products. HCl is also used in chemical synthesis. It can also be used in hydrolyzing samples for chromatographic analysis, such as in matairesinol, in flax seed, glycopeptides, κ-carrageenan derived oligosaccharides, and syn- and anti-1,3-diols. HCl is also used as a bleaching agent to neutralize alkaline agents in the food, textile, metal, and rubber industries.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

High efficiency gene transfer using chitosan/DNA nanoparticles with specific combinations of molecular weight and degree of deacetylation.
Lavertu M, et al.
Biomaterials, 27(27), 4815-4824 (2006)
Incorporation of the elderberry anthocyanins by endothelial cells increases protection against oxidative stress.
Youdim KA, et al.
Free radical biology & medicine, 29(1), 51-60 (2000)
Structural analysis of photocross linkable methacryloyl-modified protein derivatives.
Yue K, et al.
Biomaterials, 139(1), 163-171 (2017)
Marta Kolodziejczak et al.
ACS chemical neuroscience, 6(7), 1219-1230 (2015-04-11)
Maturation of functional neuronal circuits during central nervous system development relies on sophisticated mechanisms. First, axonal and dendritic growth should reach appropriate targets for correct synapse elaboration. Second, pruning and neuronal death are required to eliminate redundant or inappropriate neuronal
Douglas M McMillan et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(7), 1804-1812 (2015-01-30)
CYP2D metabolically activates codeine to morphine, which is required for codeine analgesia. Permeability across the blood-brain barrier, and active efflux, suggests that initial morphine in the brain after codeine is due to brain CYP2D metabolism. Human CYP2D is higher in

Protocols

Preparation from Packaged Powder, Preparation from Basal Salt Solutions, Banana Powder Preparation and Use, Coconut Water Preparation and Use

Follow our procedure for the determination of a chymotrypsin activity. This enzymatic assay of alpha chymotrypsin guides you through the entire process and necessary calculations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service