Merck
  • The Role of Charge Density and Hydrophobicity on the Biocidal Properties of Self-Protonable Polymeric Materials.
The Role of Charge Density and Hydrophobicity on the Biocidal Properties of Self-Protonable Polymeric Materials.
Macromolecular bioscience (2015-03-18)
Simona Matrella, Carmela Vitiello, Massimo Mella, Giovanni Vigliotta, Lorella Izzo
ABSTRACT
Intrinsic antimicrobial thermoplastic A(BC)n copolymers (n = 1, 2, 4), where A was poly(ethylene glycol) (PEG), BC was a random chain of methylmethacrylate (MMA), and alkyl-aminoethyl methacrylate (AAEMA), were synthesized and the antimicrobial activity and hemolyticity were evaluated on plaques obtained by casting as a function of the architecture, the N-substituent groups of the AAEMAs (methyl, ethyl, isopropyl, and tert-butyl groups) and the hydrophobic/charge density balance. Antimicrobial effectiveness and efficiency is controlled by the surface charge density and by the influence of N-alkyl groups on the surface morphology. Also interestingly, it is the absence of hemolitytic activity in all copolymers. In presence of Escherichia coli, the A(BC)2 copolymer with 40% of N-methyl groups is the most efficient, killing 91% of the bacteria already after 1.5 h.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Benzaldehyde dimethyl acetal, ≥98%, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Benzaldehyde dimethyl acetal, 99%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Benzaldehyde dimethyl acetal, 95%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ≥99% (calc. to H2O free subst.)
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG