Merck
  • Serum flecainide S/R ratio reflects the CYP2D6 genotype and changes in CYP2D6 activity.
Serum flecainide S/R ratio reflects the CYP2D6 genotype and changes in CYP2D6 activity.
Drug metabolism and pharmacokinetics (2015-07-22)
Kosuke Doki, Yukio Sekiguchi, Keisuke Kuga, Kazutaka Aonuma, Masato Homma
ABSTRACT
The aims of this study were to clarify whether the ratio of S- to R-flecainide (S/R ratio) in the serum flecainide concentration was associated with the stereoselectivity of flecainide metabolism, and to investigate the effects of the cytochrome P450 (CYP) 2D6 (CYP2D6) genotype and CYP2D6 inhibitor on the serum flecainide S/R ratio. In vitro studies using human liver microsomes and cDNA-expressed CYP isoforms suggested that variability in the serum flecainide S/R ratio was associated with the stereoselectivity of CYP2D6-mediated flecainide metabolism. We examined the serum flecainide S/R ratio in 143 patients with supraventricular tachyarrhythmia. The S/R ratio was significantly lower in intermediate metabolizers and poor metabolizers (IMs/PMs) than in extensive metabolizers (EMs) identified by the CYP2D6 genotype. The cut-off value for the S/R ratio to allow the discrimination between CYP2D6 EMs and IMs/PMs was 0.99. The S/R ratio in patients with co-administration of bepridil, a potent CYP2D6 inhibitor, was lower than 0.99, regardless of the CYP2D6 genotype status. Other factors, including age, sex, body weight, and renal function, did not affect the serum flecainide S/R ratio. This study suggests that the serum flecainide S/R ratio reflects the CYP2D6 genotype and changes in CYP2D6 activity on co-administration of a CYP2D6 inhibitor.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Potassium hexafluorophosphate, ≥99%
Sigma-Aldrich
Potassium hexafluorophosphate, 99.5% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Bepridil hydrochloride, powder
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Flecainide acetate salt
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Quinidine, anhydrous