Merck
Identification of a nonsense mutation in feline ABCB1.
Journal of veterinary pharmacology and therapeutics (2015-02-11)
K L Mealey, N S Burke
ABSTRACT
The aim of this study was to sequence all exons of the ABCB1 (MDR1) gene in cats that had experienced adverse reactions to P-glycoprotein substrate drugs (phenotyped cats). Eight phenotyped cats were included in the study consisting of eight cats that experienced central nervous system toxicosis after receiving ivermectin (n = 2), a combination product containing moxidectin and imidacloprid (n = 3), a combination product containing praziquantel and emodepside (n = 1) or selamectin (n = 2), and 1 cat that received the product containing praziquantel and emodepside but did not experience toxicity (n = 1). Fifteen exons contained polymorphisms and twelve exons showed no variation from the reference sequence. The most significant finding was a nonsense mutation (ABCB11930_1931del TC) in one of the ivermectin-treated cats. This cat was homozygous for the deletion mutation. All of the other phenotyped cats were homozygous for the wild-type allele. However, 14 missense mutations were identified in one or more phenotyped cats. ABCB11930_1931del TC was also identified in four nonphenotyped cats (one homozygous and three heterozygous for the mutant allele). Cats affected by ABCB11930_1931del TC would be expected to have a similar phenotype as dogs with the previously characterized ABCB1-1Δ mutation.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
REDTaq® Genomic DNA Polymerase, without MgCl2
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%