Merck
  • Direct electrochemistry of hemoglobin and biosensing for hydrogen peroxide using a film containing silver nanoparticles and poly(amidoamine) dendrimer.
Direct electrochemistry of hemoglobin and biosensing for hydrogen peroxide using a film containing silver nanoparticles and poly(amidoamine) dendrimer.
Materials science & engineering. C, Materials for biological applications (2015-10-20)
Marina Baccarin, Bruno C Janegitz, Rodrigo Berté, Fernando Campanhã Vicentini, Craig E Banks, Orlando Fatibello-Filho, Valtencir Zucolotto
ABSTRACT
A new architecture for a biosensor is proposed using a glassy carbon electrode (GCE) modified with hemoglobin (Hb) and silver nanoparticles (AgNPs) encapsulated in poly(amidoamine) dendrimer (PAMAM). The biosensors were characterized using ultraviolet-visible spectroscopy, ζ-potential and cyclic voltammetry to investigate the interactions between Hb, AgNPs and the PAMAM film. The biosensor exhibited a well-defined cathodic peak attributed to reduction of the Fe(3+) present in the heme group in Hb, as revealed by cyclic voltammetry in the presence of O2. An apparent heterogeneous electron transfer rate of 4.1s(-1) was obtained. The Hb-AgNPs-PAMAM/GCE third generation biosensor was applied in the amperometric determination of hydrogen peroxide over the linear range from 6.0 × 10(-6) to 9.1 × 10(-5)mol L(-1) with a detection limit of 4.9 × 1 0(-6)mol L(-1). The proposed method can be extended to immobilize and evaluate the direct electron transfer of other redox enzymes.
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)