Merck
  • A general approach to the synthesis and detailed characterization of magnetic ferrite nanocubes.
A general approach to the synthesis and detailed characterization of magnetic ferrite nanocubes.
Nanoscale (2015-07-08)
Yaolin Xu, Jennifer Sherwood, Ying Qin, Robert A Holler, Yuping Bao
ABSTRACT
A general approach to the synthesis and detailed characterization of magnetic ferrite nanocubes were reported, where the nanocubes were synthesized by the thermal decomposition of metal-oleate complexes following a step-heating method. The doping ions were introduced during the precursor preparation by forming M(2+)/Fe(3+) oleate mixed complex (M(2+) = Fe(2+), Mn(2+), Zn(2+), Cu(2+), Ca(2+), and Mg(2+)). The mechanistic studies showed that the presence of sodium oleate in combination with step-heating was critical for the formation of the cubic shapes for the doped magnetic ferrites. The nanocubes were extensively characterized, including morphology and crytsal structure by advanced transmission electron microscopy, doping level and distribution by energy dispersive X-ray spectroscopy and X-ray photoelectron spectroscopy, cation distribution within the spinel structures by Fourier transform infrared and Raman spectroscopy, and magnetic properties by alternating gradient magnetometer at room temperature.
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Trioctylphosphine oxide, technical grade, 90%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Sodium oleate, ≥82% (fatty acids), powder
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%