Merck
  • Affinity Capillary Electrophoresis for Selective Control of Electrophoretic Mobility of Sialic Acid Using Lanthanide-Hexadentate Macrocyclic Polyazacarboxylate Complexes.
Affinity Capillary Electrophoresis for Selective Control of Electrophoretic Mobility of Sialic Acid Using Lanthanide-Hexadentate Macrocyclic Polyazacarboxylate Complexes.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-11-13)
Daiki Goto, Kazuki Ouchi, Masami Shibukawa, Shingo Saito
ABSTRACT
It is difficult to control the electrophoretic mobility in order to obtain high resolution among saccharides in complex samples. We report herein on a new affinity capillary electrophoresis (ACE) method for an anionic monosaccharide, N-acetylneuraminic acid (Neu5Ac), which is important in terms of pathological diagnosis, using lanthanide-hexadentate macrocyclic polyazacarboxylate complexes (Ln-NOTA) as affinity reagents. It was shown that Ln-NOTA complexes increased the anionic mobility of Neu5Ac by approximately 40% through selective complexation with Neu5Ac. The extent of change in the mobility strongly depended on the type of central metal ion of Ln-NOTA. The stability constant (K) of Lu-NOTA with Neu5Ac was determined by ACE to be log Kb = 3.62 ± 0.04, which is the highest value among artificial receptors for Neu5Ac reported so far. Using this ACE, the Neu5Ac content in a glycoprotein sample, α1-acid glycoprotein (AGP), was determined after acid hydrolysis. Complete separation between Neu5Ac and hydrolysis products was successful by controlling the mobility to determine the concentration of Neu5Ac.
Product Description
Dysprosium, rod, 50mm, diameter 2.0mm, cast, 99%
Dysprosium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Dysprosium, rod, 50mm, diameter 6.35mm, cast, 99%
Dysprosium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Dysprosium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Dysprosium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Erbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Lanthanum, pieces
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Europium, wire reel, 100mm, diameter 1.0mm, hard, 99.95%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dysprosium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Dysprosium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%