Merck
  • Redox inactive metal ion triggered N-dealkylation by an iron catalyst with dioxygen activation: a lesson from lipoxygenases.
Redox inactive metal ion triggered N-dealkylation by an iron catalyst with dioxygen activation: a lesson from lipoxygenases.
Dalton transactions (Cambridge, England : 2003) (2015-05-06)
Jisheng Zhang, Yujuan Wang, Nengchao Luo, Zhuqi Chen, Kangbing Wu, Guochuan Yin
ABSTRACT
Utilization of dioxygen as the terminal oxidant at ambient temperature is always a challenge in redox chemistry, because it is hard to oxidize a stable redox metal ion like iron(III) to its high oxidation state to initialize the catalytic cycle. Inspired by the dioxygenation and co-oxidase activity of lipoxygenases, herein, we introduce an alternative protocol to activate the sluggish iron(III) species with non-redox metal ions, which can promote its oxidizing power to facilitate substrate oxidation with dioxygen, thus initializing the catalytic cycle. In oxidations of N,N-dimethylaniline and its analogues, adding Zn(OTf)2 to the [Fe(TPA)Cl2]Cl catalyst can trigger the amine oxidation with dioxygen, whereas [Fe(TPA)Cl2]Cl alone is very sluggish. In stoichiometric oxidations, it has also been confirmed that the presence of Zn(OTf)2 can apparently improve the electron transfer capability of the [Fe(TPA)Cl2]Cl complex. Experiments using different types of substrates as trapping reagents disclosed that the iron(IV) species does not occur in the catalytic cycle, suggesting that oxidation of amines is initialized by electron transfer rather than hydrogen abstraction. Combined experiments from UV-Vis, high resolution mass spectrometry, electrochemistry, EPR and oxidation kinetics support that the improved electron transfer ability of iron(III) species originates from its interaction with added Lewis acids like Zn(2+) through a plausible chloride or OTf(-) bridge, which has promoted the redox potential of iron(III) species. The amine oxidation mechanism was also discussed based on the available data, which resembles the co-oxidase activity of lipoxygenases in oxidative dealkylation of xenobiotic metabolisms where an external electron donor is not essential for dioxygen activation.
Sigma-Aldrich
1,4-Cyclohexadiene, 97%
Sigma-Aldrich
Thioanisole, ReagentPlus®, ≥99%
Sigma-Aldrich
Diphenylmethane, 99%
Sigma-Aldrich
N-Methylaniline, ≥99%
Sigma-Aldrich
Sodium trifluoromethanesulfonate, 98%
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
N,N-Dimethylaniline, purified by redistillation, ≥99.5%
Sigma-Aldrich
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
Sigma-Aldrich
N-Methylformanilide, 99%
Sigma-Aldrich
4-Bromo-N,N-dimethylaniline, 97%
Sigma-Aldrich
N-Methylaniline, 98%
Sigma-Aldrich
N,N-Dimethylaniline, ReagentPlus®, 99%
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
Copper(II) trifluoromethanesulfonate, 98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis