Merck
Interleukin-34 sustains inflammatory pathways in the gut.
Clinical science (London, England : 1979) (2015-03-25)
Eleonora Franzè, Ivan Monteleone, Maria Laura Cupi, Pamela Mancia, Flavio Caprioli, Irene Marafini, Alfredo Colantoni, Angela Ortenzi, Federica Laudisi, Giuseppe Sica, PierPaolo Sileri, Francesco Pallone, Giovanni Monteleone
ABSTRACT
IBD (inflammatory bowel disease)-related tissue damage occurs in areas which are massively infiltrated with monocytes/macrophages. These cells respond to inflammatory stimuli with enhanced production of cytokines/chemokines. In the present study, we analysed the expression and role of IL (interleukin)-34, a regulator of monocyte/macrophage differentiation, survival and function, in IBD. A significant increase in IL-34 mRNA and protein expression was seen in inflamed mucosa of patients with CD (Crohn's disease) and patients with UC (ulcerative colitis) compared with the uninvolved areas of the same patients and normal controls. IL-34 was up-regulated in LPMCs (lamina propria mononuclear cells) isolated from normal colon by TNF-α (tumour necrosis factor α) and TLR (Toll-like receptor) ligands and was down-regulated in intestinal biopsies and LPMCs of IBD patients upon treatment with infliximab. Treatment of normal LPMCs with IL-34 increased TNF-α expression in an ERK1/2 (extracellular-signal-regulated kinase 1/2)-dependent fashion and neutralization of IL-34 in IBD mucosal explants reduced TNF-α and IL-6 synthesis. In conclusion, our results indicate that IL-34 is up-regulated in IBD and suggest a role for this cytokine in sustaining the inflammatory responses in this disease.
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
IL-34 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Pricing and availability is not currently available.