Merck
Textile-based sampling for potentiometric determination of ions.
Analytica chimica acta (2015-05-24)
Grzegorz Lisak, Thomas Arnebrant, Tautgirdas Ruzgas, Johan Bobacka
ABSTRACT
Potentiometric sensing utilizing textile-based micro-volume sampling was applied and evaluated for the determination of clinically (Na(+), K(+), Cl(-)) and environmentally (Cd(2+), Pb(2+) and pH) relevant analytes. In this technological design, calibration solutions and samples were absorbed into textiles while the potentiometric cells (ion-selective electrodes and reference electrode) were pressed against the textile. Once the liquid, by wicking action, reached the place where the potentiometric cell was pressed onto the textile, hence closing the electric circuit, the potentiometric response was obtained. Cotton, polyamide, polyester and their blends with elastane were applied for micro-volume sampling. The textiles were found to influence the determination of pH in environmental samples with pH close to neutral and Pb(2+) at low analyte concentrations. On the other hand, textile-based micro-volume sampling was successfully applied in measurements of Na(+) using solid-contact sodium-selective electrodes utilizing all the investigated textiles for sampling. It was found that in order to extend the application of textile-based sampling toward environmental analysis of ions it will be necessary to tailor the physio-chemical properties of the textile materials. In general, textile-based sampling opens new possibilities for direct chemical analysis of small-volume samples and provide a simple and low-cost method to screen various textiles for their effects on samples to identify which textiles are the most suitable for on-body sensing.
Product Description
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, technical grade, 90%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, ≥97.0% (GC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Valinomycin, ≥99.0% (TLC)
Sigma-Aldrich
4-tert-Butylcalix[4]arenetetraacetic acid tetraethyl ester, 97%
Sigma-Aldrich
Tridodecylmethylammonium chloride, 98%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
2-Nitrophenyl octyl ether, 99%
Sigma-Aldrich
Valinomycin, ≥98% (TLC), ≥90% (HPLC)