Merck
Cell-free DNA in the urine of rats exposed to ionizing radiation.
Radiation and environmental biophysics (2015-05-04)
Serazhutdin A Abdullaev, Gulchachak M Minkabirova, Vladimir G Bezlepkin, Azhub I Gaziev
ABSTRACT
Investigation of cell-free DNA (cf-DNA) in body fluids, as a potential biomarker for assessing the effect of ionizing radiation on the organism, is of considerable interest. We investigated changes in the contents of cell-free mitochondrial DNA (cf-mtDNA) and cell-free nuclear DNA (cf-nDNA) in the urine of X-ray-exposed rats. Assays of cf-mtDNA and cf-nDNA were performed by a real-time PCR in rat urine collected before and after irradiation of animals with doses of 3 and 5 Gy. We also determined the presence of mutations in urine cf-mtDNA, as recognized by Surveyor nuclease. A sharp increase in cf-mtDNA and cf-nDNA in the urine of irradiated rats was observed within 24 h after exposure, followed by a decrease to normal levels. In all cases, the contents of cf-mtDNA fragment copies (estimated by gene tRNA) were significantly higher than those of cf-nDNA estimated by gene GAPDH. A certain portion of mutant cf-mtDNA fragments was detected in the urine of exposed rats, whereas they were absent in the urine of the same animals before irradiation. These preliminary data also suggest that the increased levels of urine cf-mtDNA and cf-nDNA may be a potential biomarker for noninvasive assessment of how the organism responds to ionizing radiation influence.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Guanidine thiocyanate, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Guanidine thiocyanate, BioReagent, for molecular biology, ≥99%
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%
Sigma-Aldrich
Guanidine thiocyanate, ≥97% (titration)
Sigma-Aldrich
Guanidine thiocyanate, for molecular biology, ≥99%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Guanidine thiocyanate, for molecular biology, free-flowing, Redi-Dri, ≥99.0%