Merck
  • Design, synthesis and characterisation of mannosylated ovalbumin lipid core peptide self-adjuvanting vaccine delivery system.
Design, synthesis and characterisation of mannosylated ovalbumin lipid core peptide self-adjuvanting vaccine delivery system.
Drug delivery and translational research (2015-03-20)
Pavla Simerska, Zyta Maria Ziora, Vincent Fagan, Daryn Goodwin, Farrah Edrous, Istvan Toth
ABSTRACT
Peptide-based vaccine delivery can be hampered by rapid peptidase activity and poor inherent immunogenicity. The self-adjuvanting lipid core peptide system (LCP) has been shown to confer improved stability and immunogenicity on peptide epitopes of group A Streptococcus, Chlamydia, hookworm, and malaria pathogens. However, various diseases, including cancer, still require targeted delivery of their vaccine candidates. For this reason, we have selected two model peptides (ovalbumin CD4(+) and/or CD8(+) T cell epitopes), and incorporated two or four copies of either epitope into our LCP vaccine. Optimised glycosylation of ovalbumin peptides yielded 46 % when microwave-assisted double coupling with 2 eq of carbohydrate derivative, activated by N,N-diisopropylethylamine and (O-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, was performed. All ovalbumin peptides were successfully synthesised and purified in 11-55 % yields by Fmoc- or Boc-chemistry using solid-phase peptide synthesis. The mannosylated ovalbumin peptides were nontoxic to human erythrocytes in haemolytic assay (<2 % haemolysis) and showed increased (up to 20-fold) stability in plasma.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
HATU, ≥98.0% (CHN)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in methylene chloride
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in THF
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O