Merck
  • The caffeic acid in aqueous extract of Tournefortia sarmentosa enhances neutrophil phagocytosis of Escherichia coli.
The caffeic acid in aqueous extract of Tournefortia sarmentosa enhances neutrophil phagocytosis of Escherichia coli.
Immunopharmacology and immunotoxicology (2014-10-15)
Mao-Liang Chen, Semon Wu, Tzung-Chieh Tsai, Lu-Kai Wang, Wei-Mou Chou, Fu-Ming Tsai
ABSTRACT
Tournefortia sarmentosa, a Chinese herbal medicine, is considered an antioxidant or a detoxicating agent. Recently T. sarmentosa has received attention for its effects on the immune response. Here we provide evidence that aqueous extract of T. sarmentosa can induce increased phagocytic uptake of Escherichia coli by differentiated HL-60 cells and by neutrophils. Our results also revealed that T. sarmentosa can inhibit E. coli survival within differentiated HL-60 cells. Furthermore, aqueous extract of T. sarmentosa has been shown to enhance cell surface Mac-1 expression and the activated AKT signaling pathway in E. coli-stimulated neutrophils. We also examined the effect of each constituents in aqueous extract of T. sarmentosa on phagocytic uptake of E. coli by differentiated HL-60 cells or neutrophils. Bacterial survival, cell surface Mac-1 expression, and AKT activation of neutrophils were also examined. Our results showed that caffeic acid is an important constituent in mediating aqueous extract of T. sarmentosa-induced phagocytic uptake. Taken together, these results suggest that aqueous extract of T. sarmentosa exerts effects that enhance inflammatory responses by improving phagocytic capability, inhibiting bacterial survival within cells, and increasing Mac-1 expression of neutrophils.
Product Description
Supelco
Salvianolic acid A, analytical standard
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Rosmarinic acid, primary reference standard
Sigma-Aldrich
Rosmarinic acid, 96%
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Salvianolic acid B, analytical standard
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Salvianolic acid B, European Pharmacopoeia (EP) Reference Standard
Rosmarinic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Sigma-Aldrich
Salvianolic acid B, ≥94% (HPLC)
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Tretinoin, European Pharmacopoeia (EP) Reference Standard