Merck
  • Development of artemether-loaded nanostructured lipid carrier (NLC) formulation for topical application.
Development of artemether-loaded nanostructured lipid carrier (NLC) formulation for topical application.
International journal of pharmaceutics (2014-10-08)
Petra O Nnamani, Steffi Hansen, Maike Windbergs, Claus-Michael Lehr
ABSTRACT
NLC topical formulation as an alternative to oral and parenteral (IM) delivery of artemether (ART), a poorly water-soluble drug was designed. A Phospholipon 85G-modified Gelucire 43/01 based NLC formulation containing 75% Transcutol was chosen from DSC studies and loaded with gradient concentration of ART (100-750 mg). ART-loaded NLCs were stable (-22 to -40 mV), polydispersed (0.4-0.7) with d90 size distribution range of 247-530 nm without microparticles up to one month of storage. The encapsulation efficiency (EE%) for ART in the NLC was concentration independent as 250 mg of ART loading achieved ∼61%. DSC confirmed molecular dispersion of ART due to low matrix crystallinity (0.028J/g). Ex vivo study showed detectable ART amounts after 20h which gradually increased over 48h achieving ∼26% cumulative amount permeated irrespective of the applied dose. This proves that ART permeates excised human epidermis, where the current formulation served as a reservoir to gradually control drug release over an extended period of time. Full thickness skin study therefore may confirm if this is a positive signal to hope for a topical delivery system of ART.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Artemether, ≥98% (HPLC)
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Sorbitan monostearate, meets FCC analytical specifications
Sigma-Aldrich
Di(ethylene glycol) ethyl ether, ≥99%
USP
Diethylene glycol monoethyl ether, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Diethylene glycol monoethyl ether, ReagentPlus®, 99%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%