Merck
  • Microextraction by packed sorbent (MEPS) to analyze catecholamines in innovative biological samples.
Microextraction by packed sorbent (MEPS) to analyze catecholamines in innovative biological samples.
Journal of pharmaceutical and biomedical analysis (2014-12-17)
Maria Addolorata Saracino, Laura Santarcangelo, Maria Augusta Raggi, Laura Mercolini
ABSTRACT
A new microextraction by packed sorbent (MEPS) procedure coupled to a high-performance liquid chromatographic method has been developed for quantitation of catecholamines (i.e. norepinephrine, epinephrine and dopamine) in innovative biological samples, namely dried plasma and urine spots. Analyses were carried out on a C18 reversed-phase column using a mobile phase composed of 2.5% methanol and 97.5% aqueous citrate buffer, containing octanesulfonic acid. Coulometric detection was used, setting the first analytical cell at -0.350 V and the second analytical cell at +0.400 V. Dried matrices were purified by means of a fast and feasible MEPS procedure, optimized on C18 sorbent and requiring only a small amount of biological sample. The availability of miniaturized procedures allowed satisfying specific requirements that ought to be considered during pre-treatment intended for catecholamine analysis. The extraction yield values were always higher than 85% and sensitivity was good, with a limit of quantitation of 100 pg mL(-1) for all the analytes. Satisfactory results were also obtained in terms of linearity, precision and accuracy. The method was successfully applied to dried plasma and urine spots derived from two socially diversified groups, namely psychiatric patients and poly-drug abusers, in comparison to healthy volunteers.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)