Merck
  • Construction of a portable sample preparation device with a magnetic poly(methacrylic acid-co-ethylene dimethacrylate) monolith as the extraction medium and its application in the enrichment of UV filters in water samples.
Construction of a portable sample preparation device with a magnetic poly(methacrylic acid-co-ethylene dimethacrylate) monolith as the extraction medium and its application in the enrichment of UV filters in water samples.
Journal of separation science (2014-07-22)
Jing Li, Li Xu, Qiong-Wei Yu, Zhi-Guo Shi, Ting Zhang, Yan Liu
ABSTRACT
A portable sample preparation device with a magnetic polymer monolith as the extraction medium was constructed. The monolith was synthesized by polymerizing methacrylic acid and ethylene dimethacrylate around a cylindrical magnet. In this way, the monolith with a magnetic core could be readily attached to the extraction device by magnetism. The constructed device was evaluated for the enrichment of UV filters in water samples, followed by high-performance liquid chromatographic analysis. The extraction efficiency for the targets was satisfactory with no matrix interference. Good linearities were obtained for the UV filters with the correlation coefficients >0.9986. The limits of detection and quantification for the UV filters were 0.3-0.8 and 1.0-2.4 ng/mL, respectively. The recoveries of the UV filters from the spiked water samples at the concentration of 100 ng/mL were 95.3-101.7%, with relative standard deviations <10%. Accordingly, the proposed portable device was demonstrated to be suitable for on-site simultaneous sampling, purification, and preconcentration within a single step.
Product Description
Supelco
Oxybenzone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2,4-Dihydroxybenzophenone, 99%
Supelco
Oxybenzone, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Supelco
Melting point standard 47-49°C, analytical standard
USP
Oxybenzone, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Hydroxybenzophenone, 98%
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Sigma-Aldrich
2-Hydroxy-4-methoxybenzophenone, 98%
Benzophenone, European Pharmacopoeia (EP) Reference Standard