Merck
  • Metrics and clinical relevance of percutaneous penetration and lateral spreading.
Metrics and clinical relevance of percutaneous penetration and lateral spreading.
Skin pharmacology and physiology (2014-09-18)
Aline Vieille-Petit, Nicholas Blickenstaff, Garrett Coman, Howard Maibach
ABSTRACT
Percutaneous penetration of urea in vivo in man has been documented. If urea can penetrate the skin, it may also move laterally. Lateral spreading of topical substances leads to unpredictable penetration dynamics and increased skin surface area exposure. The ability of urea, a low molecular-weight hydrophilic model, to penetrate the stratum corneum (SC) and spread outside the application site was investigated in vitro using tape stripping with spectroscopy. The parameters investigated were the following: time between urea application and tape stripping, formulations containing urea and use of a petrolatum-covered ring barrier around the marked application area. The percentage of urea was determined in and around the application site. The spreading of topically applied urea to neighboring areas occurred and was time but not formulation dependent. A significant difference between protocols with and without the petrolatum ring was observed. These results suggest the clinical importance of lateral spreading, occurring predominately on the skin surface. SC thickness varies between anatomical sites, predisposing areas such as the face and scalp margins to increased percutaneous penetration of topical products. The use of a protective petrolatum ring can inhibit lateral spreading of hair dye in individuals allergic to hair dye, limit systemic absorption and increase accuracy when assessing penetration dynamics.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, analytical standard
Sigma-Aldrich
Ethylene brassylate, ≥95%, FG
Supelco
2,3-Butanedione monoxime, for spectrophotometric det. of urea, ≥99.0%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications