Merck
  • Sodium L-ascorbate enhances elastic fibers deposition by fibroblasts from normal and pathologic human skin.
Sodium L-ascorbate enhances elastic fibers deposition by fibroblasts from normal and pathologic human skin.
Journal of dermatological science (2014-07-13)
Aleksander Hinek, Hyunjun J Kim, Yanting Wang, Andrew Wang, Thomas F Mitts
ABSTRACT
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic fibers in cultures of fibroblasts derived from normal human skin and dermal fat, as well as in explants of normal human skin, stretch-marked skin and keloids. Effects of SA on the extracellular matrix production were assessed quantitatively by PCR analyses, western blots, biochemical assay of insoluble elastin and by immuno-histochemistry. We also evaluated effects of SA on production of the reactive oxygen species (ROS) and phosphorylation of IGF-I and insulin receptors. SA, applied in 50-200 μM concentrations, stimulates production of both collagen and elastic fibers in all tested cultures. Moreover, combination of SA with a proline hydroxylase inhibitor induces a beneficial remodelling in explants of dermal scars, resulting in the inhibition of collagen deposition and induction of new elastogenesis. Importantly, we revealed that SA stimulates elastogenesis only after intracellular influx of non-oxidized ascorbate anions (facilitated by the sodium-dependent ascorbate transporter), that causes reduction of intracellular ROS, activation of c-Src tyrosine kinase and the enhancement of IGF-1-induced phosphorylation of the IGF-1 receptor that ultimately triggers elastogenic signalling pathway. Our results endorse the use of this potent stimulator of collagen and elastin production in the treatment of wrinkled and stretch-marked skin. They also encourage inclusion of SA into therapeutic combinations with collagenogenesis inhibitors to prevent formation of dermal scars and keloids.
Sigma-Aldrich
Propidium iodide solution
Pricing and availability is not currently available.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Pricing and availability is not currently available.
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Valine, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Valine, ReagentPlus®, ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Pricing and availability is not currently available.
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Pricing and availability is not currently available.
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Valine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.