Merck
Biomimetic and nanostructured hybrid bioactive glass.
Biomaterials (2015-03-05)
Xianfeng Zhou, Nita Sahai, Lin Qi, Steven Mankoci, Weilong Zhao
ABSTRACT
Inspired by nature's toughening mechanisms, we designed a new polyhedral oligomeric silsesquioxane (POSS)-derived hybrid glass (PHG) that has covalent interactions on the molecular scale between the inorganic POSS cage and organic phase. These features allow "elastic deformation" of the inorganic POSS cage in limited scale. The final product is a bulk hybrid material with toughness (3.56 ± 0.25 MPa·m(1/2)) similar to natural bone (2.4-5.3 MPa·m(1/2)). PHG exhibited excellent bioactivity by promoting the formation of plate-like hydroxyapatite on its surface in simulated body fluid and showed good cell adhesion. PHG also can be a platform to guide adipose tissue-derived mesenchymal stem cells differentiation and mineralization. The key structural features of this material can be used to guide the design of bio-inspired composites with unique toughness, which would be of great benefit to hard tissue engineering.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Triethoxysilane, 95%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
PSS-Octavinyl substituted
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
o-Xylene, anhydrous, 97%