Skip to Content
Merck
All Photos(1)

Key Documents

442719

Supelco

Nonanal

analytical standard

Synonym(s):

Aldehyde C9, Nonyl aldehyde, Pelargonaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CHO
CAS Number:
Molecular Weight:
142.24
Beilstein:
1236701
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

~0.26 mmHg ( 25 °C)

Assay

≥99.5% (GC)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.424 (lit.)

bp

93 °C/23 mmHg (lit.)

density

0.827 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCC=O

InChI

1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h9H,2-8H2,1H3

InChI key

GYHFUZHODSMOHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ke-Ming Li et al.
Journal of chemical ecology, 38(3), 287-294 (2012-03-10)
Odorant receptors are thought to play critical roles in the perception of chemosensory stimuli by insects. The primary method to address the functions of odorant receptors in insects is to use in vitro binding assays between the receptors and potential
Thomas Van Hecke et al.
PloS one, 9(6), e101122-e101122 (2014-07-01)
The effects of fat content and nitrite-curing of pork were investigated on the formation of cytotoxic and genotoxic lipid oxidation products (malondialdehyde, 4-hydroxy-2-nonenal, volatile simple aldehydes), protein oxidation products (protein carbonyl compounds) and NOC-specific DNA adducts (O6-carboxy-methylguanine) during in vitro
Dirk Louis P Schorkopf et al.
The Journal of experimental biology, 212(Pt 8), 1153-1162 (2009-03-31)
Like ants and termites some species of stingless bees (Meliponini), which are very important pollinators in the tropics, use pheromone trails to communicate the location of a food source. We present data on the communicative role of mandibular gland secretions
Carmine Summo et al.
Meat science, 86(3), 839-844 (2010-08-04)
An experimental investigation was carried out to study the evolution of the volatile compounds of ripened sausages as a function of both storage time (time) and composition of packaging atmosphere (MAP). Ripened sausages, prepared at an industrial plant, were packed
Elena Chiarpotto et al.
BioFactors (Oxford, England), 24(1-4), 229-236 (2006-01-13)
Transient activation of fibroblasts or fibroblast-like cells to proliferate and to produce elevated quantities of extracellular matrix is essential to fibrosis. This activation is regulated by several cytokines produced by various inflammation-associated cells. Among these, transforming growth factor beta1 (TGFbeta1)

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service