Merck
  • Effects of food and gender on the pharmacokinetics of ginkgolides A, B, C and bilobalide in rats after oral dosing with ginkgo terpene lactones extract.
Effects of food and gender on the pharmacokinetics of ginkgolides A, B, C and bilobalide in rats after oral dosing with ginkgo terpene lactones extract.
Journal of pharmaceutical and biomedical analysis (2014-08-29)
Ping Huang, Liang Zhang, Chuan Chai, Xiao-Cui Qian, Wen Li, Jun-Song Li, Liu-Qing Di, Bao-Chang Cai
ABSTRACT
The ginkgo terpene lactones (GTL), mainly including bilobalide (BB), ginkgolide A (GA), ginkgolide B (GB) and ginkgolide C (GC) possess different biological activities such as peripheral vasoregulation, platelet-activating factor (PAF) receptor antagonism, neuroprotective properties and prevention of membrane damage caused by free radicals. To investigate the effects of food and gender on the bioavailability of BB, GA, GB and GC after oral administration of GTL extract, a rapid UPLC-MS/MS method was developed and validated. A reversed phase C18 column (100mm×2.1mm, i.d., 1.7μm) and a mobile phase consisted of methanol and 1mM ammonium acetate (70/30, v/v) were employed. Compared with the fasted group, the t1/2 values for BB, GA, GB and GC in fed were all increased (p<0.05), AUC0-t and AUC0-∞ values of BB, GA, GB and GC were all significantly increased (p<0.05), but the Cmax values of BB, GA, GB and GC were significantly decreased (p<0.05). In comparison with the male group, all of the t1/2 values and AUC0-t values for BB, GA, GB and GC in female were higher (p<0.05), but no statistical difference in Tmax values for BB, GA, GB and GC between these two groups. Food and gender factor showed significant effects on the pharmacokinetics of BB, GA, GB, and GC. The results suggested that oral doses of GTL should be lowered for fasted and female subjects, compared with the fed and male subjects, respectively.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ginkgo Biloba Terpene Lactones Mix, 100 μg/mL each component in acetonitrile, ampule of 1.0 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Methanol, analytical standard