Skip to Content
Merck
All Photos(1)

Key Documents

320390

Sigma-Aldrich

Methanol

Methanol

≥99.8%, ACS reagent

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
41116121
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
64.7 °C (lit.)
vapor pressure:
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Methanol, ACS reagent, ≥99.8%

grade

ACS reagent

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

dilution

(for analytical testing)

impurities

H2SO4, passes test (darkened)
MnO4- reducers, passes test
≤0.0002 meq/g Titr. base
≤0.0003 meq/g Titr. acid
≤0.001% acetaldehyde
≤0.001% acetone
≤0.001% formaldehyde
≤0.1% water

evapn. residue

≤0.001%

color

APHA: ≤10
clear

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol is a polar solvent and feedstock to produce chemicals and fuel additives. It can be produced through the catalytic hydrogenation of CO2.

Application

Methanol can be used as a solvent in the:
  • Palladium(II)-exchanged hydroxyapatite-catalyzed Suzuki–Miyaura cross-coupling reactions.
  • Reductive amination of ketones with ammonia and dihydrogen to synthesize primary amines.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of digestive enzyme activity in larvae of spotted sand bass Paralabrax maculatofasciatus II: electrophoretic analysis.
Alvarez-Gonzalez CA, et al.
Fish Physiology and Biochemistry, 36(1), 29-37 (2010)
Carotenoid Extraction and Quantification from Capsicum annuum.
Richins RD, et al.
Bio-protocol, 4(19), e1256-e1256 (2014)
The conversion of methanol and other O-compounds to hydrocarbons over zeolite catalysts.
Chang CD and Silvestri AJ.
J. Catal., 47(2), 249-259 (1977)
Electrocatalysis by ad-atoms: Part II. Enhancement of the oxidation of methanol on platinum by ruthenium ad-atoms.
Watanabe MA and Motoo S.
J. Electroanal. Chem. Interfac. Electrochem., 60(3), 267-273 (1975)
Timo Heidt et al.
Circulation research, 115(2), 284-295 (2014-05-03)
Macrophages populate the steady-state myocardium. Previously, all macrophages were thought to arise from monocytes; however, it emerged that, in several organs, tissue-resident macrophages may self-maintain through local proliferation. Our aim was to study the contribution of monocytes to cardiac-resident macrophages

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service