Merck
  • Physicochemical and immunological assessment of engineered pure protein particles with different redox states.
Physicochemical and immunological assessment of engineered pure protein particles with different redox states.
ACS nano (2015-02-26)
Katelyn T Gause, Yan Yan, Jiwei Cui, Neil M O'Brien-Simpson, Jason C Lenzo, Eric C Reynolds, Frank Caruso
ABSTRACT
The development of subunit antigen delivery formulations has become an important research endeavor, especially in cases where a whole cell vaccine approach has significant biosafety issues. Particle-based systems have shown particular efficacy due to their inherent immunogenicity. In some cases, fabrication techniques can lead to changes in the redox states of encapsulated protein antigens. By employing a uniform, well-characterized, single-protein system, it is possible to elucidate how the molecular details of particle-based protein antigens affect their induced immune responses. Using mesoporous silica-templated, amide bond-stabilized ovalbumin particles, three types of particles were fabricated from native, reduced, and oxidized ovalbumin, resulting in particles with different physicochemical properties and immunogenicity. Phagocytosis, transcription factor activation, and cytokine secretion by a mouse macrophage cell line did not reveal significant differences between the three types of particles. Oxidation of the ovalbumin, however, was shown to inhibit the intracellular degradation of the particles compared with native and reduced ovalbumin particles. Slow intracellular degradation of the oxidized particles was correlated with inefficient antigen presentation and insignificant levels of T cell priming and antibody production in vivo. In contrast, particles fabricated from native and reduced ovalbumin were rapidly degraded after internalization by macrophages in vitro and resulted in significant T cell and B cell immune responses in vivo. Taken together, the current study demonstrates how the redox state of a protein antigen significantly impacts the immunogenicity of the particulate vaccine formulations.
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
MES monohydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES monohydrate, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
MOPS, BioUltra, Molecular Biology, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
SAFC
MOPS
SAFC
MES hydrate
SAFC
MES monohydrate
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride, 95% (HPLC)