Merck
  • Comprehensive quantitative analysis of purines and pyrimidines in the human malaria parasite using ion-pairing ultra-performance liquid chromatography-mass spectrometry.
Comprehensive quantitative analysis of purines and pyrimidines in the human malaria parasite using ion-pairing ultra-performance liquid chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-05)
Christian D Laourdakis, Emilio F Merino, Andrew P Neilson, Maria B Cassera
ABSTRACT
Targeted metabolite profiling has aided in the understanding of a variety of biological processes in the malaria parasite as well as in drug discovery. A fast and sensitive analytical method, based on ion-pairing reversed phase ultra-high performance liquid chromatography tandem mass spectrometry (IP-RP-UPLC-MS/MS), was optimized for the simultaneous analysis of intracellular levels of 35 purine and pyrimidine nucleobases, nucleosides, and nucleotides. This analytical method allows for chromatographic separation of highly polar metabolites using reverse phase chemistry within 15 min. The analytical performance of the method was evaluated and successfully applied to the quantification of purines and pyrimidines in Plasmodium falciparum and its host cell, the human erythrocyte. In addition, this method can be customized to include other related metabolites such as NADPH and NADP, among others.
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Uridine 5′-monophosphate, ≥98%
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenine, European Pharmacopoeia (EP) Reference Standard
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%