Merck
  • Evaluating the efficiency of isotope transmission for improved panel design and a comparison of the detection sensitivities of mass cytometer instruments.
Evaluating the efficiency of isotope transmission for improved panel design and a comparison of the detection sensitivities of mass cytometer instruments.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2015-02-24)
Sabine Tricot, Mickael Meyrand, Chiara Sammicheli, Jamila Elhmouzi-Younes, Aurélien Corneau, Sylvie Bertholet, Marie Malissen, Roger Le Grand, Sandra Nuti, Hervé Luche, Antonio Cosma
ABSTRACT
The recent introduction of mass cytometry, a technique coupling a cell introduction system generating a stream of single cells with mass spectrometry, has greatly increased the number of parameters that can be measured per single cell. As with all new technology there is a need for dissemination of standardization and quality control procedures. Here, we characterize variations in sensitivity observed across the mass range of a mass cytometer, using different lanthanide tags. We observed a five-fold difference in lanthanide detection over the mass range and demonstrated that each instrument has its own sensitivity pattern. Therefore, the selection of lanthanide combinations is a key step in the establishment of a staining panel for mass cytometry-based experiments, particularly for multicenter studies. We propose the sensitivity pattern as the basis for panel design, instrument standardization and future implementation of normalization algorithms.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Dysprosium, foil, not light tested, 5x5mm, thickness 0.005mm, as rolled, 99%
Dysprosium, powder, 10g, max. particle size 500 micron, 99.9%
Dysprosium, powder, 1g, max. particle size 500 micron, 99.9%
Dysprosium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%
Dysprosium, powder, 5g, max. particle size 500 micron, 99.9%
Dysprosium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Dysprosium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Dysprosium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Dysprosium, rod, 50mm, diameter 2.0mm, cast, 99%
Dysprosium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Dysprosium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Dysprosium, rod, 100mm, diameter 6.35mm, cast, 99%
Dysprosium, rod, 50mm, diameter 6.35mm, cast, 99%