Merck
  • Antimicrobial susceptibility and mechanisms of high-level macrolide resistance in clinical isolates of Moraxella nonliquefaciens.
Antimicrobial susceptibility and mechanisms of high-level macrolide resistance in clinical isolates of Moraxella nonliquefaciens.
Journal of medical microbiology (2013-11-08)
Shotaro Nonaka, Kosuke Matsuzaki, Tomoya Kazama, Hiroyuki Nishiyama, Yoko Ida, Saho Koyano, Kazunari Sonobe, Noboru Okamura, Ryoichi Saito
ABSTRACT
We investigated antimicrobial susceptibility and the molecular mechanism involved in conferring high-level macrolide resistance in 47 clinical isolates of Moraxella nonliquefaciens from Japan. Antimicrobial susceptibility was determined using Etest and agar dilution methods. Thirty-two erythromycin-non-susceptible strains were evaluated for the possibility of clonal spreading, using PFGE. To analyse the mechanism related to macrolide resistance, mutations in the 23S rRNA gene and the ribosomal proteins, and the presence of methylase genes were investigated by PCR and sequencing. The efflux system was examined using appropriate inhibitors. Penicillin, ampicillin, amoxicillin, cefixime, levofloxacin and antimicrobials containing β-lactamase inhibitors showed strong activity against 47 M. nonliquefaciens isolates. Thirty-two (68.1 %) of the 47 isolates showed high-level MICs to macrolides (MIC ≥128 mg l(-1)) and shared the A2058T mutation in the 23S rRNA gene. The geometric mean MIC to macrolides of A2058T-mutated strains was significantly higher than that of WT strains (P<0.0001). Thirty-two isolates with high-level macrolide MICs clustered into 30 patterns on the basis of the PFGE dendrogram, indicating that the macrolide-resistant strains were not clonal. In contrast, no common mutations of the ribosomal proteins or methylase genes, or overproduction of the efflux system were observed in A2058T-mutated strains. Moreover, of the 47 M. nonliquefaciens strains, 43 (91.5 %) were bro-1 and 4 (8.5 %) were bro-2 positive. Our results suggest that most M. nonliquefaciens clinical isolates show high-level macrolide resistance conferred by the A2058T mutation in the 23S rRNA gene. This study represents the first characterization of M. nonliquefaciens.
Sigma-Aldrich
Azithromycin
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Azithromycin, European Pharmacopoeia (EP) Reference Standard
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Josamycin, ≥90% (HPLC)
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
USP
Azithromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
USP
Azithromycin Identity, United States Pharmacopeia (USP) Reference Standard
Azithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Josamycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Josamycin, European Pharmacopoeia (EP) Reference Standard