Skip to Content
Merck
All Photos(1)

Documents

1046056

USP

Azithromycin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Azithromycin dihydrate, 1-Oxa-6-azacyclopentadecan-15-one, 13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-alpha-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10- trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-beta-D-xylohexopyranosyl]oxy]-, dihydrate, [2R-(2R, 3S, 4R, 5R, 8R, 10R, 11R, 12S, 13S, 14R)]-, N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A, CP-62993

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H72N2O12 · 2H2O
CAS Number:
Molecular Weight:
785.02
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

USP

API family

azithromycin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C38H72N2O12.2H2O/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28;;/h20-33,35,41-43,45-46H,15-19H2,1-14H3;2*1H2/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-;;/m1../s1

InChI key

SRMPHJKQVUDLQE-KUJJYQHYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azithromycin is a subclass of macrolides consisting of 15-membered macrocyclic lactone ring to which sugar moieties are linked.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Azithromycin USP reference standard for use in specified quality tests and assays.

Also used to prepare standard, standard stock solutions for the assay, identification, performance, and impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Azithromycin for Oral Suspension
  • Azithromycin Capsules
  • Azithromycin for Injection

Biochem/physiol Actions

Azithromycin dihydrate is a macrolide antibiotic, azalide subclass. It binds to the 50S subunit of the 70S bacterial ribosomes and inhibits RNA-dependent protein synthesis in bacterial cells. Azithromycin also has anti-immunomodulatory/anti-inflammatory properties, which make it useful in treating cystic fibrosis.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here
Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azithromycin Capsules
United States Pharmacopeia, 27(6), 450-450 (2013)
Azithromycin for Injection
United States Pharmacopeia, 44(2), 451-451 (2020)
Azithromycin for Oral Suspension
United States Pharmacopeia, 44(5) (2022)
Aspects of degradation kinetics of azithromycin in aqueous solution.
Zhang, Yu, et al.
Chromatographia, 70.1-2, 67-73 (2009)
Development and validation of a micellar liquid chromatographic method with UV detection for determination of azithromycin in tablets and capsules.
Kulikov, A. U., and A. G. Verushkin.
Chromatographia, 60.1-2, 33-38 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service