Merck
  • Fascaplysin induces caspase mediated crosstalk between apoptosis and autophagy through the inhibition of PI3K/AKT/mTOR signaling cascade in human leukemia HL-60 cells.
Fascaplysin induces caspase mediated crosstalk between apoptosis and autophagy through the inhibition of PI3K/AKT/mTOR signaling cascade in human leukemia HL-60 cells.
Journal of cellular biochemistry (2015-01-07)
Suresh Kumar, Santosh Kumar Guru, Anup Singh Pathania, Sudhakar Manda, Ajay Kumar, Sandip B Bharate, Ram A Vishwakarma, Fayaz Malik, Shashi Bhushan
ABSTRACT
In this study, we for the first time explored the cellular and molecular mechanism of anticancer properties of fascaplysin, a marine sponge-derived alkaloid. Our study demonstrated that fascaplysin induced a cooperative interaction between apoptotic and autophagic pathways to induce cytotoxicity in HL-60 cells. Fascaplysin treatment not only activated pro-apoptotic events like PARP-1 cleavage and caspase activation but also triggered autophagy signaling as shown by the increased expression of LC3-II, ATG7and beclin. Interestingly, it was found that use of pan-caspase inhibitor completely reversed the fascaplysin mediated cell death as analyzed by MTT and cell cycle assays. It was observed that cell death as well as the expression of pro-death proteins was partially reversed, when key autophagy mediators ATG7 was silenced by siRNA in fascaplysin treated cells. Cooperative involvement of autophagy and apoptotic signaling in cytotoxicity was confirmed when combined silencing of pro-apototic (PARP-1) and autophagic (ATG-7) signaling by respective siRNA's lead to substantial rescue of cell death induced by fascaplysin. Although, apoptosis and autophagy are two independent cell death pathways, our findings provide detailed insight by which both the pathways acted cooperatively to elicit fascaplysin induced cell death in HL-60 cells. Our findings provide molecular insight into the anti-cancer potential of fascaplysin by showing that both autophagic and apoptotic signaling can work together in the induction of cell death.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Di(ethylene glycol) ethyl ether, ≥99%
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sigma-Aldrich
Propidium iodide solution
SAFC
L-Glutamine
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
USP
Diethylene glycol monoethyl ether, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Rhodamine 123, BioReagent, ≥85% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethylene glycol monoethyl ether, ReagentPlus®, 99%