Skip to Content
Merck
All Photos(1)

Documents

W297003

Sigma-Aldrich

Pyruvic acid

≥97%, FG

Synonym(s):

α-Ketopropionic acid, 2-Oxopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOOH
CAS Number:
Molecular Weight:
88.06
FEMA Number:
2970
Beilstein:
506211
EC Number:
Council of Europe no.:
19
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39021320
PubChem Substance ID:
Flavis number:
8.019
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.428 (lit.)

bp

165 °C (lit.)

mp

11-12 °C (lit.)

density

1.267 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; acidic; sour

storage temp.

2-8°C

SMILES string

CC(=O)C(O)=O

InChI

1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)

InChI key

LCTONWCANYUPML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Asparagus, beef, sake, Virginia tobacco, wheat bread.
Pyruvic acid is the key component formed during the hydrolysis of flavor-precursors called S-alk(en)yl-L-cysteine-sulfoxides in onion tissues by allinase during maceration or chopping. The amount of pyruvic acid formed is used as a measure for onion pungency.

Biochem/physiol Actions

Taste at 5 ppm

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simplified pyruvic acid analysis suitable for onion breeding programs.
Yoo KS, et al.
Hortscience: a Publication of the American Society For Horticultural Science Hortscience, 30(6), 1306-1306 (1995)
Development of an automated system for pyruvic acid analysis in onion breeding.
Yoo KS & Pike LM.
Sci. Hortic., 82(3), 193-201 (1999)
Onion flavor and odor, enzymatic development of pyruvic acid in onion as a measure of pungency.
Schwimmer S & Weston WJ
Journal of Agricultural and Food Chemistry, 9(4), 301-304 (1961)
Emilie A Bard-Chapeau et al.
Nature genetics, 46(1), 24-32 (2013-12-10)
The most common risk factor for developing hepatocellular carcinoma (HCC) is chronic infection with hepatitis B virus (HBV). To better understand the evolutionary forces driving HCC, we performed a near-saturating transposon mutagenesis screen in a mouse HBV model of HCC.
Sarah C Atkinson et al.
Plant molecular biology, 81(4-5), 431-446 (2013-01-29)
Lysine is one of the most limiting amino acids in plants and its biosynthesis is carefully regulated through inhibition of the first committed step in the pathway catalyzed by dihydrodipicolinate synthase (DHDPS). This is mediated via a feedback mechanism involving

Protocols

Science Slam panel: Leading gene therapy developers discuss commercialization challenges and the importance of robust process development plans.

Science Slam panel: Leading gene therapy developers discuss commercialization challenges and the importance of robust process development plans.

Science Slam panel: Leading gene therapy developers discuss commercialization challenges and the importance of robust process development plans.

Science Slam panel: Leading gene therapy developers discuss commercialization challenges and the importance of robust process development plans.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service