Merck
  • Evaluation of amino acid ester-based ionic liquids as buffer additives in CE for the separation of 2-arylpropionic acids nonsteroidal anti-inflammatory drugs.
Evaluation of amino acid ester-based ionic liquids as buffer additives in CE for the separation of 2-arylpropionic acids nonsteroidal anti-inflammatory drugs.
Electrophoresis (2014-05-24)
Maria C Mavroudi, Constantina P Kapnissi-Christodoulou
ABSTRACT
The aim of the present study is the CE performance evaluation for the separation of 2-arylpropionic acid nonsteroidal anti-inflammatory drugs. In particular, the separation of indoprofen, carprofen, ketoprofen, ibuprofen, and flurbiprofen was obtained by supporting the BGE either with SDS or an amino acid ester-based ionic liquid (AAIL). The performance of these additives was evaluated by comparing migration times, efficiencies and %RSD values. The addition of the AAIL into the BGE provided baseline separation within 10 min, while in the case of SDS, the analytes eluted within 23 min. The optimum conditions involve a BGE of 100 mM Tris/10 mM sodium tetraboratedecahydrate (pH 8) and 40 mM l-alanine tert butyl ester lactate or 10 mM SDS and a temperature of 35°C for AAIL and 20°C for SDS. The run-to-run reproducibility was evaluated by computing the %RSD values of the EOF and the analyte peaks. When the AAIL was used, an excellent reproducibility was obtained, since all %RSD values were below 1.3%. On the contrary, the addition of SDS resulted in much higher RSD values (2.1-11.7%). The efficiency values of all analyte peaks were above 102 000 for l-AlaC4 Lac, in comparison to SDS, which provided efficiency values between 47000 and 76000. Finally, in an attempt to study the synergistic effect of SDS and AAIL, both additives were added into the BGE at concentrations of 10 and 40 mM, respectively. The results were similar to the ones obtained when SDS was used as the sole additive.
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
2-Chloro-2-methylpropane, 99%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioXtra, ≥99.5% (T)
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Supelco
Ibuprofen
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, puriss. p.a., ACS reagent, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Carprofen for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Chloro-2-methylpropane, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis