Merck
  • The synthetic triterpenoid (CDDO-Im) inhibits STAT3, as well as IL-17, and improves DSS-induced colitis in mice.
The synthetic triterpenoid (CDDO-Im) inhibits STAT3, as well as IL-17, and improves DSS-induced colitis in mice.
Inflammopharmacology (2014-04-10)
Leo R Fitzpatrick, Elizabeth Stonesifer, Jeffrey S Small, Karen T Liby
ABSTRACT
Synthetic triterpenoids inhibit IL-17 and improve autoimmune disease in mice. A prototype triterpenoid, 1-[2-cyano-3-,12-dioxooleana-1,9(11)-dien-28-oyl] imidazole (CDDO-Im), also inhibits signal transducer and activator of transcription 3 (STAT3) activation. The goals of our study were twofold: (1) To determine if ex vivo treatment with CDDO-Im attenuated colonic IL-17 secretion from isolated splenocytes and colonic strips; (2) To determine if oral treatment with CDDO-Im improved DSS-induced colitis in mice. Splenocytes were isolated from male Balb/c mice. Colitis was induced in rodents, with either trinitrobenzene sulfonic acid or dextran sulfate sodium (DSS). Colonic strips were collected 5 or 6 days after colitis induction. Splenocytes or colonic strips were exposed to CDDO-Im (0.5-2 μM) concomitantly with IL-23 + IL-1β. Supernatants were collected after 48 or 24 h, and IL-17 was measured by ELISA. Using a DSS colitis model, mice were dosed orally with vehicle or CDDO-Im (20 mg/kg) over a 5-day period. Subsequently, various parameters of colitis were determined on study day 6. Ex vivo treatment with CDDO-Im inhibited IL-17 secretion from splenocytes and colonic strips. The IC50 values were ≤0.62 μM. In vivo, CDDO-Im improved the altered colonic histology, and cytokine (IL-6, and IL-17) contents. Colonic STAT3 activation was also significantly reduced by CDDO-Im treatment. CDDO-Im attenuated IL-17 secretion in ex vivo models of inflammation. In vivo, histological and biochemical parameters of colitis were improved in CDDO-Im treated mice. CDDO-Im has a unique pharmacological profile, which supports further testing in animal models of IBD.
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Human IL-17 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)