Merck
  • Reduction of nitroaromatics sorbed to black carbon by direct reaction with sorbed sulfides.
Reduction of nitroaromatics sorbed to black carbon by direct reaction with sorbed sulfides.
Environmental science & technology (2015-02-12)
Wenqing Xu, Joseph J Pignatello, William A Mitch
ABSTRACT
Sorption to black carbons is an important sink for organic contaminants in sediments. Previous research has suggested that black carbons (graphite, activated carbon, and biochar) mediate the degradation of nitrated compounds by sulfides by at least two different pathways: reduction involving electron transfer from sulfides through conductive carbon regions to the target contaminant (nitroglycerin) and degradation by sulfur-based intermediates formed by sulfide oxidation (RDX). In this study, we evaluated the applicability of black carbon-mediated reactions to a wider variety of contaminant structures, including nitrated and halogenated aromatic compounds, halogenated heterocyclic aromatic compounds, and halogenated alkanes. Among these compounds, black carbon-mediated transformation by sulfides over a 3-day time scale was limited to nitroaromatic compounds. The reaction for a series of substituted nitroaromatics proceeded by reduction, as indicated by formation of 3-bromoaniline from 3-bromonitrobenzene, and inverse correlation of log kobs with energy of the lowest unoccupied molecular orbital (ELUMO). The log kobs was correlated with sorbed sulfide concentration, but no reduction of 3-bromonitrobenzene was observed in the presence of graphite and sulfite, thiosulfate, or polysulfides. Whereas nitroglycerin reduction occurred in an electrochemical cell containing sheet graphite electrodes in which the reagents were placed in separate compartments, nitroaromatic reduction only occurred when sulfides were present in the same compartment. The results suggest that black carbon-mediated reduction of sorbed nitroaromatics by sulfides involves electron transfer directly from sorbed sulfides rather than transfer of electrons through conductive carbon regions. The existence of three different reaction pathways suggests a complexity to the sulfide-carbon system compared to the iron-carbon system, where contaminants are reduced by electron transfer through conductive carbon regions.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
1-Bromo-3-nitrobenzene, 97%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.