Merck
  • Pseudokinase tribbles 1 (TRB1) negatively regulates tumor-suppressor activity of p53 through p53 deacetylation.
Pseudokinase tribbles 1 (TRB1) negatively regulates tumor-suppressor activity of p53 through p53 deacetylation.
Biological & pharmaceutical bulletin (2015-04-04)
Chiharu Miyajima, Yasumichi Inoue, Hidetoshi Hayashi
ABSTRACT
Tribbles 1 (TRB1) is one of the mammalian orthologs of Drosophila Tribbles, which regulates development and cell proliferation. TRB1 is suggested to act as a scaffold protein in signaling pathways for important cellular processes. TRB1 has also been identified as a myeloid oncogenic driver and mediates leukemogenesis through the mitogen-activated protein extracellular kinase (MEK)/extracellular signal-regulated kinase (ERK) pathway and CCAAT/enhancer binding protein (C/EBP) transcriptional factors. However, the physiological roles of TRB1 in solid tumors have not been clarified. Here, we show that TRB1 interacts with p53 and suppresses its tumor suppressor activity. TRB1 knockdown enhances transcriptional activity of p53 and decreases cell viability. Interestingly, TRB1 enhances histone deacety lase 1 (HDAC1)-mediated p53 deacetylation and decreases DNA binding of p53. These results suggest that TRB1 is involved in the proliferation of tumor cells by inhibiting the activities of tumor suppressor p53 in solid tumors.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Anti-Tribbles-1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
7-Amino-1,3-naphthalenedisulfonic acid monopotassium salt monohydrate, BioReagent, suitable for fluorescence, ≥98.0% (T)