Skip to Content
Merck
All Photos(1)

Key Documents

09-126

Sigma-Aldrich

Anti-Tribbles-1 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRIB1(10221)

General description

The Tribbles family of proteins are kinase-like proteins that contain a kinase-like domain, but lack either a catalytic domain or a protein-protein interaction domain (2). They are believed to regulate signal processing for various pathways including MAPK, Akt, and NFkB that are involved in various aspects of development, diabetes, and cancer (1,2,3). Their involvement has been reported to regulate a number of pathways that include mitosis, cell activation, and apoptosis. They are are a family made up of three family members. Tribbles-1 (TRIB1, SKIP1, G-protein-coupled receptor induced protein, and phosphoprotein regulated by mitogenic pathways) is a modulator of MAPK action. It has been shown to directly interact with the upstream kinases MEK1 and MKK4 of Erk and JNK, respectively(1). Tribble-2 (Trib2, TRB2) has been shown to interact with Akt(2). Tribbles-3 (TRIB3, TRB3. NIPK, SKIP3) has been shown to interact with Akt and the transcription factors ATF4 and NFkB p65 (RelA). It is interaction with RelA is thought to be negative regulation. Interestingly, it has been reported to regulate Akt activation via insulin(3).

Specificity

Tribbles-1

Immunogen

KLH-conjugated sythetic linear peptide directed against a region of the N-terminal portion of Tribbles-1.

Application

This Anti-Tribbles-1 Antibody is validated for use in IH, WB for the detection of Tribbles-1.

Quality

routinely evaluated by western blot on lysate from HeLa cells

Target description

41 kDa

Physical form

100 μg in 100μL of affinity purified rabbit serum in storage buffer containing PBS with 0.05% sodium azide.

Analysis Note

Control
HeLa

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaori Tsuzuki et al.
FEBS letters, 593(3), 369-380 (2018-12-18)
Tribbles related homolog 1 is the mammalian ortholog of Tribbles, which controls cell division and migration during development in Drosophila. TRB1 is a pseudokinase and functions as a scaffold protein. Recent findings suggest that TRB1 plays important roles in hepatic
Jessica M Johnston et al.
Science advances, 5(10), eaax9183-eaax9183 (2019-11-07)
Macrophages drive atherosclerotic plaque progression and rupture; hence, attenuating their atherosclerosis-inducing properties holds promise for reducing coronary heart disease (CHD). Recent studies in mouse models have demonstrated that Tribbles 1 (Trib1) regulates macrophage phenotype and shows that Trib1 deficiency increases
Hye Youn Sung et al.
Biology, 1(1), 43-57 (2012-01-01)
Development of the atherosclerotic plaque involves a complex interplay between a number of cell types and an extensive inter-cellular communication via cell bound as well as soluble mediators. The family of tribbles proteins has recently been identified as novel controllers
Chiharu Miyajima et al.
Biological & pharmaceutical bulletin, 38(4), 618-624 (2015-04-04)
Tribbles 1 (TRB1) is one of the mammalian orthologs of Drosophila Tribbles, which regulates development and cell proliferation. TRB1 is suggested to act as a scaffold protein in signaling pathways for important cellular processes. TRB1 has also been identified as
Chiharu Miyajima et al.
Biological & pharmaceutical bulletin, 38(8), 1126-1133 (2015-08-04)
Tribbles 1 (TRB1), a member of the Tribbles family, is a pseudokinase that is conserved among species and implicated in various human diseases including leukemia, cardiovascular diseases, and metabolic disorders. However, the role of TRB1 in the immune response is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service